Learn More
Invitrogen™ GBP2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA544204
Product Code. 15576392
  • € 518.00 / 100µL
Description
Peptide sequence: HTKGIWMWCV PHPKKPEHTL VLLDTEGLGD IEKGDNENDS WIFALAILLS Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GBP2
Polyclonal
Unconjugated
GBP2
GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67
Rabbit
Affinity Chromatography
RUO
2634
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P32456
GBP2
Synthetic peptide directed towards the N-terminal of human GBP2 (aa 74-123).