missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP5 Antibody (OTI5C9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72343G
This item is not returnable.
View return policy
Description
GBP5 Monoclonal specifically detects GBP5 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| GBP5 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GBP5 | |
| Full length recombinant protein of human GBP5 (NP_443174) produced in HEK293T cells. | |
| 0.1 mL | |
| 115362.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI5C9 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| GBP-5, GBP-TA antigen, GTP-binding protein 5, Guanine nucleotide-binding protein 5, guanylate binding protein 5, guanylate-binding protein 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction