missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP6 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97207V
This item is not returnable.
View return policy
Description
GBP6 Polyclonal antibody specifically detects GBP6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GBP6 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| DKFZp686G0786, FLJ39135, GBP-6, GTP-binding protein 6, Guanine nucleotide-binding protein 6, guanylate binding protein family, member 6, guanylate-binding protein 6 | |
| Produced in rabbits immunized with E. coli-derived Human GBP6 fragment. | |
| 0.1 mL | |
| Innate Immunity | |
| 163351 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction