missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCAP2 Antibody (A1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-5422DL594
This item is not returnable.
View return policy
Description
GCAP2 Monoclonal antibody specifically detects GCAP2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GCAP2 | |
| Monoclonal | |
| DyLight 594 | |
| DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48 | |
| Bacterial expressed full length GCAP-2. | |
| 0.1 mL | |
| Signal Transduction, Vision | |
| 2979 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| A1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction