missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCDH, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00002639-D01P.100ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Sequence: MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASKTekniske data
| GCDH | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_000159.2 | |
| GCDH | |
| GCDH (NP_000150.1, 1 a.a. ∼ 438 a.a) full-length human protein. | |
| RUO | |
| 2639 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| glutaryl-Coenzyme A dehydrogenase | |
| ACAD5/GCD | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion