missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCFC1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71922B
This item is not returnable.
View return policy
Description
GCFC1 Polyclonal antibody specifically detects GCFC1 in Human samples. It is validated for Western Blot
Specifications
| GCFC1 | |
| Polyclonal | |
| Western Blot | |
| BM020, C21orf66, chromosome 21 open reading frame 66, FLJ90561, fSAP105, functional spliceosome-associated protein 105, GCFCFSAP105, GC-rich sequence DNA-binding factor 1, GC-rich sequence DNA-binding factor candidate | |
| The immunogen for this product maps to a region between residue 275 and 325 of human GC-rich Sequence DNA-binding Factor 1 using the numbering given in entry NP_057715.2 (GeneID 94104). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 94104 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction