missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GCHFR Antibody (GCHFR/7734) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26940H
This item is not returnable.
View return policy
Description
GCHFR Monoclonal antibody specifically detects GCHFR in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GCHFR | |
| Monoclonal | |
| HRP | |
| GFRPP35, GTP cyclohydrolase I feedback regulator, GTP cyclohydrolase I feedback regulatory proteinGTP cyclohydrolase 1 feedback regulatory protein, HsT16933, MGC138467, MGC138469, p35 | |
| Recombinant fragment human GCHFR protein (exact sequence is proprietary) | |
| 0.1 mL | |
| metabolism | |
| 2644 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| GCHFR/7734 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction