missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCKR Polyclonal Antibody

Brand: Thermo Scientific PA5115274
This item is not returnable.
View return policy
Description
Specifications
| GCKR | |
| Polyclonal | |
| Unconjugated | |
| Gckr | |
| FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 231103, 25658, 2646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q07071, Q14397, Q91X44 | |
| Gckr | |
| A synthesized peptide derived from human GCKR(Accession Q14397), corresponding to amino acid residues G207-S257. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction