missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCLC Antibody (OTI1A3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70830PE
This item is not returnable.
View return policy
Description
GCLC Monoclonal antibody specifically detects GCLC in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| GCLC | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1A3 | |
| Western Blot, Immunohistochemistry | |
| EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit | |
| Full length human recombinant protein of human GCLC(NP_001489) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology, Lipid and Metabolism | |
| 2729 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction