missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCLM Polyclonal Antibody
GREENER_CHOICE

Product Code. 16329804
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16329804 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16329804 Supplier Invitrogen™ Supplier No. PA582843

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RTLHLQTGNL LNWGRLRKKC PSTHSEELHD CIQKTLNEWS SQINPDLVRE FPDVLECTVS HAVE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCLM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gclm
Gene Accession No. P48507
Gene Alias AI649393; gamma glutamylcysteine synthetase; gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; Gclm; Gcmc; GCS light chain; Glclr; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; glutamate cysteine ligase, modifier subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate--cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase, modifier subunit; GSC light chain; null
Gene Symbols Gclm
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCLM. Recombinant protein control fragment (Product #RP-93984).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.