Learn More
GCM1 Monoclonal Antibody (OTI1F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810539
This item is not returnable.
View return policy
Description
This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.Specifications
| GCM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP62 | |
| GCM1 | |
| Full length human recombit protein of human GCM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI1F4 | |
| Unconjugated | |
| GCM1 | |
| GCMA, hGCMa | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8521 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |