missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCM1 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810524
This item is not returnable.
View return policy
Description
This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.Specifications
| GCM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP62 | |
| GCM1 | |
| Full length human recombit protein of human GCM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5A5 | |
| Unconjugated | |
| GCM1 | |
| GCMA, hGCMa | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8521 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction