Learn More
GCN1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-97851
Product Code. 18739903
  • € 583.00 / 0.10mL
Description
GCN1L1 Polyclonal specifically detects GCN1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Store at 4C. Do not freeze.