missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN4 Antibody (C11L34), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81274AF647
This item is not returnable.
View return policy
Description
GCN4 Monoclonal specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| GCN4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| GCN4 | |
| Biotinylated GCN4-peptide (HLENEVARLKK). | |
| 0.1 mL | |
| Primary | |
| This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity. | |
| Store at 4°C in the dark. | |
| IgG λ |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| C11L34 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 856709 | |
| Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur