missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCNF/NR6A1 Antibody (OTI6F3), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70825C
This item is not returnable.
View return policy
Description
GCNF/NR6A1 Monoclonal specifically detects GCNF/NR6A1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GCNF/NR6A1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR6A1 | |
| Recombinant protein fragment corresponding to amino acids 1-248 of human GCNF/NR6A1 (NP_001480) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI6F3 | |
| Western Blot | |
| GCNF1, GCNFNR61, Germ cell nuclear factor, hGCNF, nuclear receptor subfamily 6 group A member 1, nuclear receptor subfamily 6, group A, member 1, retinoic acid receptor-related testis-associated receptor, Retinoid receptor-related testis-specific receptor, RTRhRTR | |
| Mouse | |
| Affinity Purified | |
| GPCR, Olfactory | |
| 2649.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction