missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCR2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97163G
This item is not returnable.
View return policy
Description
GCR2 Polyclonal specifically detects GCR2 in Human samples. It is validated for Western Blot.
Specifications
| GCR2 | |
| Polyclonal | |
| Western Blot | |
| ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X | |
| Produced in rabbits immunized with E. coli-derived Human GCR2 fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 11319 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction