missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46215
This item is not returnable.
View return policy
Description
GCR2 Polyclonal specifically detects GCR2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GCR2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| ECD | |
| The immunogen for this product maps to a region between residue 614 and 644 of human Suppressor of GCR2 using the numbering given in entry NP_009196.1 (GeneID 11319). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 11319 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction