missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDAP1L1 Antibody (OTI1G5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72182PE
This item is not returnable.
View return policy
Description
GDAP1L1 Monoclonal specifically detects GDAP1L1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| GDAP1L1 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| GDAP1L1 | |
| Full length recombinant protein of human GDAP1L1 (NP_076939) produced in HEK293T cells. | |
| 0.1 mL | |
| 78997.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1G5 | |
| Flow Cytometry | |
| dJ881L22.1, dJ995J12.1.1, DKFZp761K228, ganglioside-induced differentiation-associated protein 1-like 1, GDAP1-L1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction