Learn More
GDAP1L1 Mouse anti-Human, Clone: OTI1A5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503224
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.Specifications
| GDAP1L1 | |
| Monoclonal | |
| Unconjugated | |
| GDAP1L1 | |
| dJ881L22.1, dJ995J12.1.1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 78997 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1A5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96MZ0 | |
| GDAP1L1 | |
| Full length human recombit protein of human GDAP1L1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.