missing translation for 'onlineSavingsMsg'
Learn More

GDAP1L1 Mouse anti-Human, Clone: OTI1G5, TrueMAB™, Origene

Product Code. 17528661
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17528661 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17528661 Supplier Origene Technologies Supplier No. TA503153S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDAP1L1 Monoclonal antibody specifically detects GDAP1L1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen GDAP1L1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1G5
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GDAP1L1
Gene Accession No. Q96MZ0
Gene Alias dJ881L22.1; dJ995J12.1.1; ganglioside induced differentiation associated protein 1 like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; Gdap1l; GDAP1L1; GDAP1-L1
Host Species Mouse
Immunogen Full length human recombinant protein of human GDAP1L1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78997
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.