missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDAP1L1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15898033
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15898033 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898033 Supplier Invitrogen™ Supplier No. PA569628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The ganglioside GD3 synthase causes cell differentiation with neurite sprouting when transfected into the mouse neuroblastoma cell line Neuro2a. After differentiation, the expression of several genes is upregulated, including one that encodes a protein termed ganglioside-induced differentiation-associated protein 1 (Gdap1). A similar gene was found in humans, and mutations in the human gene are associated with Charcot-Marie-Tooth type 4A disease. The protein encoded by this gene is similar in sequence to the human GDAP1 protein. Several transcript variants encoding different isoforms, as well as a noncoding transcript variant, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDAP1L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GDAP1L1
Gene Accession No. 0
Gene Alias dJ881L22.1; dJ995J12.1.1; ganglioside induced differentiation associated protein 1 like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside induced differentiation associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; ganglioside-induced differentiation-associated protein 1-like 1; Gdap1l; GDAP1L1; GDAP1-L1
Gene Symbols GDAP1L1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat GDAP1L1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311616
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.