missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDC-0152, MedChemExpress
Description
GDC-0152 is a potent IAPs inhibitor, and binds to the BIR3 domains of XIAP, cIAP1, cIAP2 and the BIR domain of ML-IAP with Ki values of 28 nM, 17 nM, 43 nM and 14 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | GDC-0152 |
| CAS | 873652-48-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H34N6O3S |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (200.55 mM) ∣Ethanol : 50 mg/mL (100.27 mM; Need ultrasonic) |
| SMILES | O=C(N1[C@H](C(NC2=C(C3=CC=CC=C3)N=NS2)=O)CCC1)[C@H](C4CCCCC4)NC([C@H](C)NC)=O |
| Molecular Weight (g/mol) | 498.64 |
| Formula Weight | 498.64 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?