missing translation for 'onlineSavingsMsg'
Learn More

GDC-0152, MedChemExpress

Product Code. 16485788
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485788 10 mg 10mg
16411947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485788 Supplier Medchem Express Supplier No. HY13638

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0152 is a potent IAPs inhibitor, and binds to the BIR3 domains of XIAP, cIAP1, cIAP2 and the BIR domain of ML-IAP with Ki values of 28 nM, 17 nM, 43 nM and 14 nM, respectively.

GDC-0152 is a potent IAPs inhibitor, and binds to the BIR3 domains of XIAP, cIAP1, cIAP2 and the BIR domain of ML-IAP with Ki values of 28 nM, 17 nM, 43 nM and 14 nM, respectively.

Specifications

Chemical Name or Material GDC-0152
CAS 873652-48-3
Color White
Purity Grade Notes Research
Molecular Formula C25H34N6O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (200.55 mM) ∣Ethanol : 50 mg/mL (100.27 mM; Need ultrasonic)
SMILES O=C(N1[C@H](C(NC2=C(C3=CC=CC=C3)N=NS2)=O)CCC1)[C@H](C4CCCCC4)NC([C@H](C)NC)=O
Molecular Weight (g/mol) 498.64
Formula Weight 498.64
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.