missing translation for 'onlineSavingsMsg'
Learn More

GDC-0326, MedChemExpress

Product Code. 16434566
Encompass
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434566

Brand: Medchem Express HY101272

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress GDC-0326 is a potent and selective PI3Kα inhibitor with a Ki of 0.2 nM.

GDC-0326 is a potent and selective PI3Kα inhibitor with a Ki of 0.2 nM.

Specifications

Chemical Name or Material GDC-0326
CAS 1282514-88-8
Color White
Purity Grade Notes Research
Molecular Formula C19H22N6O3
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (261.49 mM)
SMILES C[C@H](OC1=CC=C2C3=NC(C4=NC=NN4C(C)C)=CN3CCOC2=C1)C(N)=O
Molecular Weight (g/mol) 382.42
Formula Weight 382.42
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.