missing translation for 'onlineSavingsMsg'
Learn More

GDC-0339, MedChemExpress

Product Code. p-200097587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427238 10 mg 10mg
16409767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16427238 Supplier Medchem Express Supplier No. HY16976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0339 is a potent, orally bioavailable and well tolerated pan-Pim kinase inhibitor, with Kis of 0.03 nM, 0.1 nM and 0.02 nM for Pim1, Pim2 and Pim3, respectively. GDC-0339 is discovered as a potential treatment of multiple myeloma.

GDC-0339 is a potent, orally bioavailable and well tolerated pan-Pim kinase inhibitor, with Kis of 0.03 nM, 0.1 nM and 0.02 nM for Pim1, Pim2 and Pim3, respectively. GDC-0339 is discovered as a potential treatment of multiple myeloma.

Specifications

Chemical Name or Material GDC-0339
CAS 1428569-85-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22F3N7OS
Quantity 10 mg
Solubility Information DMSO : ≥ 52 mg/mL (111.71 mM)
SMILES FC1=CC=CC(F)=C1C2=NC(C(NC3=C(N4CC[C@@H](F)[C@H](N)CC4)N(C)N=C3)=O)=C(N)S2
Molecular Weight (g/mol) 465.5
Formula Weight 465.5
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.