missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-15 Antibody (CL0328), Novus Biologicals™
Mouse Monoclonal Antibody
€ 302.00 - € 554.00
Specifications
| Antigen | GDF-15 |
|---|---|
| Clone | CL0328 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686118
|
Novus Biologicals
NBP2-52917-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685026
|
Novus Biologicals
NBP2-52917 |
0.1 mL |
€ 554.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-15 Monoclonal antibody specifically detects GDF-15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| GDF-15 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LSLAEASRASFPGPSELHSEDSRFRELRKRYEDLLTRLRANQSWEDSNTDLVPAPAVRILTPEVRLGSGGHLHLRISRAALPEGLPEASRLHRALFRLSPTASRSWDVTRPLRRQLSLARPQAPALHLRLSPPPSQSDQL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL0328 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Mouse | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 9518 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title