missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-15 Antibody (MM0307-10A3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12331FR
This item is not returnable.
View return policy
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| GDF-15 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GDF15 | |
| Human recombinant GDF-15 | |
| 0.1 mL | |
| Primary | |
| This antibody was selected for its ability to detect human GDF-15. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Flow Cytometry | |
| MM0307-10A3 | |
| Western Blot, Flow Cytometry | |
| growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
| Mouse | |
| Protein G purified | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| 9518 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction