missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 498.00
Specifications
| Antigen | GDF-15 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637446
|
Novus Biologicals
NBP2-68911-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672807
|
Novus Biologicals
NBP2-68911 |
100 μg |
€ 498.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-15 Polyclonal antibody specifically detects GDF-15 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| GDF-15 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol | |
| 9518 | |
| IgG | |
| Protein A purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EVQVTMCIGACPSQFRAANMHAQIKTSLHRLKPDTVPAPCCVPASYNPMVLIQKTDTGVSLQTYDDLLAKDCHC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title