missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98686UV
This item is not returnable.
View return policy
Description
GDF-3 Polyclonal antibody specifically detects GDF-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GDF-3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GDF-3 | |
| 0.1 mL | |
| Cytokine Research, Stem Cell Markers | |
| 9573 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction