missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-3 Antibody (SR1219), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | GDF-3 |
|---|---|
| Clone | SR1219 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677094
|
Novus Biologicals
NBP3-22018-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654034
|
Novus Biologicals
NBP3-22018-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-3 Monoclonal antibody specifically detects GDF-3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GDF-3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6 | |
| A synthesized peptide derived from human GDF-3 (Uniprot #: Q9NR23) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1219 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9573 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title