missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-6/BMP-13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76364-0.025mg
This item is not returnable.
View return policy
Description
GDF-6/BMP-13 Polyclonal specifically detects GDF-6/BMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| GDF-6/BMP-13 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BMP13, CDMP2, GDF16, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| NP_001001557 | |
| GDF6 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human GDF6. The immunogen is located within amino acids 320 - 370 of GDF6. Amino Acid Squence: AFASRHGKRHGKKSRLR | |
| 0.025 mg | |
| Vision | |
| 392255 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction