missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-8/Myostatin Antibody (GDF9/4261), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08680AF405
This item is not returnable.
View return policy
Description
GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA
Specifications
| GDF-8/Myostatin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
| Recognizes the 113 amino acid carboxy-terminal fragment of GDF-8/Myostatin protein (Uniprot: O14793) | |
| 100 μg | |
| Cytokine Research | |
| 2660 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| GDF9/4261 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction