missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-8/Myostatin Antibody (RM0243-13F2), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12334MFV450
This item is not returnable.
View return policy
Description
GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GDF-8/Myostatin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0243-13F2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
| Mouse recombinant GDF-8/Myostatin | |
| 0.1 mL | |
| Cytokine Research | |
| 2660 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction