Learn More
GDF-9 Antibody (72B), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61933PE
Product Code. 18615727
  • € 575.00 / 0.10mL
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, ELISA
Rat
0.1 mL
Cytokine Research
Purified
PBS
Protein G purified
RUO
Store at 4C in the dark.