Learn More
GDF-9 Antibody (72B), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-61933PE
This item is not returnable.
View return policy
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Western Blot, ELISA | |
| Rat | |
| 0.1 mL | |
| Cytokine Research | |
| Purified |
| PBS | |
| Protein G purified | |
| RUO | |
| Store at 4C in the dark. | |
| IgG1 |