missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566356
This item is not returnable.
View return policy
Description
Immunogen sequence: DFQGDALQPED FLEEDEYHAT TETVISMAQE TDPAVQTDGS PLCCHFHFSP KVMF Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.
Specifications
| GDF11 | |
| Polyclonal | |
| Unconjugated | |
| GDF11 | |
| BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10220 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95390 | |
| GDF11 | |
| Recombinant Human GDF11. Recombinant protein control fragment (Product #RP-107036). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction