missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103058
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GDF3.
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Specifications
| GDF3 | |
| Polyclonal | |
| Unconjugated | |
| GDF3 | |
| C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14562, 500311, 9573 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07104, Q9NR23 | |
| GDF3 | |
| A synthesized peptide derived from human GDF3(Accession Q9NR23), corresponding to amino acid residues F167-Q217. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction