missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI1 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810977S
This item is not returnable.
View return policy
Description
GDI1 Monoclonal antibody specifically detects GDI1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| GDI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3C5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26994 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B1 | |
| Unconjugated | |
| RNF11 | |
| 1A; GDI1; GDI-1; GDIA; GDIalpha; GDIL; GDP dissociation inhibitor 1; GDP-dissociation inhibitor 1; GTP-binding protein; guanosine diphosphate (GDP) dissociation inhibitor 1; guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; MRX41; MRX48; oligophrenin-2; OPHN2; Protein XAP-4; rab GDI alpha; Rab GDIalpha; rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; RABGD1A; RABGDIA; SMG p25A GDI; testis secretory sperm-binding protein Li 208a; XAP4; XAP-4 | |
| Human recombinant protein fragment corresponding to amino acids 2-154 of human RNF11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction