missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI1 Monoclonal Antibody (OTI7F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810981
This item is not returnable.
View return policy
Description
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.Specifications
| GDI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P31150 | |
| GDI1 | |
| Full length human recombit protein of human GDI1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7F7 | |
| Unconjugated | |
| GDI1 | |
| 1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2664 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction