missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 156.00 - € 478.00
Specifications
| Antigen | GDI1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17780544
![]() |
Invitrogen™
PA5119136 |
200 μL |
€ 478.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17783354
|
Invitrogen™
PA5121960 |
20 μL |
€ 156.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDI1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI1 is expressed primarily in neural and sensory tissues. Mutations in GDI1 have been linked to X-linked nonspecific mental retardation.Specifications
| GDI1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Pig | |
| GDI1 | |
| 1A; GDI1; GDI-1; GDIA; GDIalpha; GDIL; GDP dissociation inhibitor 1; GDP-dissociation inhibitor 1; GTP-binding protein; guanosine diphosphate (GDP) dissociation inhibitor 1; guanosine diphosphate dissociation inhibitor 1; mental retardation, X-linked 48; MRX41; MRX48; oligophrenin-2; OPHN2; Protein XAP-4; rab GDI alpha; Rab GDIalpha; rab GDP dissociation inhibitor alpha; rab GDP-dissociation inhibitor, alpha; RABGD RABGDIA; RABGD1A; RABGDIA; SMG p25A GDI; testis secretory sperm-binding protein Li 208a; XAP4; XAP-4 | |
| GDI1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P31150, P50396 | |
| 100623607, 14567, 2664 | |
| Recombinant protein GDP Dissociation Inhibitor 1. The antigen corresponds to amino acid range 1-274 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title