missing translation for 'onlineSavingsMsg'
Learn More

GDI2 Antibody (0B10) [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30520155 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520155 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520155 Supplier Novus Biologicals Supplier No. NBP342973AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDI2 Monoclonal antibody specifically detects GDI2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDI2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 0B10
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias beta, FLJ16452, GDP dissociation inhibitor 2, Guanosine diphosphate dissociation inhibitor 2, RABGDIBFLJ37352
Host Species Mouse
Immunogen GDP dissociation inhibitor 2 - (Uniprot# P50395)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2665
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.