missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDNF Antibody (MM0308-12F9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-12335CL1
This item is not returnable.
View return policy
Description
GDNF Monoclonal antibody specifically detects GDNF in Human samples. It is validated for ELISA,Western Blot,Neutralization
Specifications
| GDNF | |
| Monoclonal | |
| CoraFluor 1 | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
| Human recombinant GDNF | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 2668 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| ELISA, Western Blot, Neutralization | |
| MM0308-12F9 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction