missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDNF Antibody (MM0308-12F9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12335JF646
This item is not returnable.
View return policy
Description
GDNF Monoclonal antibody specifically detects GDNF in Human samples. It is validated for Western Blot
Specifications
| GDNF | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MM0308-12F9 | |
| Western Blot | |
| Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
| Human recombinant GDNF | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 2668 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction