Learn More
Fisherbrand™ Gel Casting Gate
Click to view available options
For Use With (Equipment):
Maxi Plus Standard
Maxi Standard
Unit Size:
1 pair
Each
Description
- Casting gates
- Integral silicone seals effectively seal the tray without the need for tape
Specifications
Specifications
| For Use With (Equipment) | Maxi Plus Standard |
| Type | Gel Casting Gate |
| Includes | Integral Silicone Seals |