missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU20 Mini-gel Horizontal Electrophoresis Unit

Offers complete control over loading volume and well depth.

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11749635

Quantity Price
1 € 73.84 / Each
10 € 64.14 / Each
Estimated Shipment
17-03-2021
Add to basket


Description

Description

  • Color coded
  • Height adjustable
  • For use with HU20 Mini-gel Horizontal Electrophoresis Unit
Specifications

Specifications

HU20 Mini-gel Horizontal Electrophoresis Unit
28
1 mm
5 mm
Comb
1 Each
20 μL
Videos
SDS
Documents