missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GEM Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142886
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
GEM belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction.The protein encoded by this gene belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
| GEM | |
| Polyclonal | |
| Unconjugated | |
| GEM | |
| AV020497; Gem; GTP binding protein (gene overexpressed in skeletal muscle); GTP binding protein overexpressed in skeletal muscle; GTP-binding mitogen-induced T-cell protein; GTP-binding protein GEM; GTP-binding protein overexpressed in skeletal muscle; kinase-inducible Ras-like protein; KIR; MGC26294; RAS-like protein KIR | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P55040 | |
| GEM | |
| Peptide with sequence C-QKEPHQYSHRNRH (aa 34-46). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction