Learn More
GEMC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41131
 View more versions of this product
Product Code. 18184809
  • € 532.00 / 0.10mg
Description
GEMC1 Polyclonal specifically detects GEMC1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GEMC1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
GMNC
Antibody was raised against a 19 amino acid synthetic peptide near the center of human GEMC1. The immunogen is located within amino acids 130 - 180 of GEMC1. Amino Acid Squence: YGKFRKGKRKSKEQRYSPA
0.1 mg
Primary
Human, Mouse
IgG
Western Blot, ELISA
1 mg/mL
Western Blot 1 μg/mL, ELISA
GEMC1, Geminin Coiled-Coil Domain Containing, Geminin Coiled-Coil Domain-Containing Protein 1, GMNC
Rabbit
Affinity Purified
RUO
647309
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.