missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41131-0.025mg
This item is not returnable.
View return policy
Description
GEMC1 Polyclonal specifically detects GEMC1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| GEMC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GMNC | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human GEMC1. The immunogen is located within amino acids 130 - 180 of GEMC1. Amino Acid Squence: YGKFRKGKRKSKEQRYSPA | |
| 0.025 mg | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA | |
| GEMC1, Geminin Coiled-Coil Domain Containing, Geminin Coiled-Coil Domain-Containing Protein 1, GMNC | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 647309 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction