Learn More
Thermo Scientific Chemicals Gemfibrozil, 98%
Product Code. p-7112898 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15713457
Brand: Thermo Scientific Chemicals 461790050
Request Bulk or Custom Format
CAS: 25812-30-0 | C15H22O3 | 250.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25812-30-0
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00079335
InChI Key HEMJJKBWTPKOJG-UHFFFAOYSA-N
Synonym gemfibrozil, lopid, 5-2,5-dimethylphenoxy-2,2-dimethylpentanoic acid, jezil, decrelip, lipur, bolutol, apo-gemfibrozil, gemfibromax, cholespid
PubChem CID 3463
ChEBI CHEBI:5296
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1
Specifications
Melting Point 60.0°C to 61.0°C
Quantity 5 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 250.33
Percent Purity 98%
Chemical Name or Material Gemfibrozil