missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gemin 6 Monoclonal Antibody (3D2)
GREENER_CHOICE

Product Code. 15589142
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15589142 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15589142 Supplier Invitrogen™ Supplier No. MA522604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MSEWMKKGPL EWQDYIYKEV RVTASEKNEY KGWVLTTDPV SANIVLVNFL EDGSMSVTGI MGHAVQTVET MNEGDHRVRE KLMHLFTSGD CKAYSPEDLE ERKNSLKKWL EKNHIPITEQ GDAPRTLCVA GVLTIDPPYD PENCSSSNEI ILSRVQDLIE GHLTASQ.

GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GEMIN6
Gene Accession No. Q8WXD5
Gene Alias 2610019B15Rik; 2810470M17Rik; FLJ23459; gem (nuclear organelle) associated protein 6; gem nuclear organelle associated protein 6; gem-associated protein 6; gemin 6; Gemin6; gemin-6; SIP2
Gene Symbols GEMIN6
Host Species Mouse
Immunogen GEMIN6 (AAH18195, 1 a.a. ~ 167 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79833
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.