missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77177H
This item is not returnable.
View return policy
Description
GEMIN2 Polyclonal antibody specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| GEMIN2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SIP1. The immunogen is located within the last 50 amino acids of SIP1. Amino Acid Squence: VSRYFDQRDLADEPS | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8487 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction